Ethylacetat
Glasflasche, 1 L
Katalognr. ROTH6784.3
Marketsource
Anbieter: Roth Carl
Direkt zu
Product Details & Documents
Synonyme: Acetic acid ethyl ester
- CAS Nummer:141-78-6
- Formel:CH₃COOC₂H₅
- MDL-Nummer:MFCD00009171
- Molekulargewicht:88.11 g/mol
- Merck-Index:13,03792
- Siedepunkt:77,1 °C (1013 hPa)
- Schmelzender Platingehalt:–83 °C
- Dichte:0,902 g/cm³ (20 °C)
- Blitz Pt:–4 °C
- Lagertemperatur:Raumtemperatur
- EINECS:205-500-4
- UN:1173
- ADR:3,II
- ERREICHEN:01-2119475103-46
Technische Daten
Technische Daten
- Packung:Glasflasche
- VE:1 L
- Cat. No.:ROTH6784.3
- Umweltzertifizierung:Chem21 Solvent Selection Guide recommended by default and after discussion
- Umweltfreundlichkeit:Unbedenklicher für Mensch und Umwelt
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



