Nitrobenzol
Glasflasche, 500 mL
Catalog # ROTH4394.2
Marketsource
Supplier: Roth Carl
Jump to:
Product Details & Documents
- CAS Number:98-95-3
- Formula:C₆H₅NO₂
- MDL Number:MFCD00007043
- Molecular Weight:123.11 g/mol
- Merck Index:13,06621
- Boiling Pt:210,8 °C (1013 hPa)
- Melting Pt:5,7 °C
- Density:1,2037 g/cm³ (20 °C)
- Flash Pt:88 °C
- Storage Temperature:Raumtemperatur
- EINECS:202-716-0
- UN:1662
- ADR:6.1,II
Specifications
Specifications
- Packung:Glasflasche
- VE:500 mL



