Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
 

L(+)-Leucin

Glass bottle for solids, 250 g
Catalog # ROTH3984.3
Marketsource
Supplier:   Roth Carl
L(+)-Leucin
Catalog # ROTH3984.3
Supplier:  Roth Carl
Marketsource
CAS Number:  61-90-5
L(+)-Leucin

Glass bottle for solids, 250 g

CAS Number:  61-90-5
Price
115.80 SFr.
Log in for availability
1 * 250 g
Product Details & Documents
Synonyms:  (S)-(+)-Leucine, Leu, H-Leu-OH, Leucine, L-Leucine
  • CAS Number:61-90-5
  • Formula:C₆H₁₃NO₂
  • MDL Number:MFCD00002617
  • Molecular Weight:131.17 g/mol
  • Merck Index:14,05451
  • Melting Pt:300 °C
  • Density:1,165 g/cm³ (20 °C)
  • Storage Temperature:Raumtemperatur
  • EINECS:200-522-0
Specifications

Specifications

  • Packung:
    Glass bottle for solids
  • VE:
    250 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.