L(+)-Leucin
Glass bottle for solids, 1 kg
ROTH3984.2
Marketsource
: (S)-(+)-Leucine, Leu, H-Leu-OH, Leucine, L-Leucine
- :61-90-5
- :C₆H₁₃NO₂
- :MFCD00002617
- :131.17 g/mol
- :14,05451
- :300 °C
- :1,165 g/cm³ (20 °C)
- :Raumtemperatur
- :200-522-0
- Packung:Glass bottle for solids
- VE:1 kg