L(+)-Leucin
Glass bottle for solids, 100 g
Katalognr. ROTH3984.1
Marketsource
Anbieter: Roth Carl
Direkt zu
Product Details & Documents
Synonyme: (S)-(+)-Leucine, Leu, H-Leu-OH, Leucine, L-Leucine
- CAS Nummer:61-90-5
- Formel:C₆H₁₃NO₂
- MDL-Nummer:MFCD00002617
- Molekulargewicht:131.17 g/mol
- Merck-Index:14,05451
- Schmelzender Platingehalt:300 °C
- Dichte:1,165 g/cm³ (20 °C)
- Lagertemperatur:Raumtemperatur
- EINECS:200-522-0
Technische Daten
Technische Daten
- Packung:Glass bottle for solids
- VE:100 g
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



