Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Natriumchlorid
Natriumchlorid
5 kg
  ROTH3957.2
Marketsource
 :   Roth Carl
Natriumchlorid
  ROTH3957.2
 :  Roth Carl
Marketsource
 :  7647-14-5
Natriumchlorid
5 kg
 :  7647-14-5
SFr. 94.20
1 * 5 kg
 :  Saline, NaCl, Sea salt, Halite
  •  :7647-14-5
  •  :NaCl
  •  :MFCD00003477
  •  :58.44 g/mol
  •  :14,08599
  •  :1413 °C (1013 hPa)
  •  :801 °C
  •  :2,16 g/cm³ (20 °C)
  •  :Raumtemperatur
  •  :231-598-3
  • VE:
    5 kg
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.