Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
 

Multiple distributors for bottles, BOLA

3 × max. 8,5 mm, 3 × GL 14
Katalognr. BOHLD613-08
Anbieter:   Bohlender
Multiple distributors for bottles, BOLA
Katalognr. BOHLD613-08
Anbieter:  Bohlender
Multiple distributors for bottles, BOLA
Multiple distributors for bottles, BOLA

3 × max. 8,5 mm, 3 × GL 14

Preis
130.00 SFr.
Einloggen für Verfügbarkeit
1 * 1 ST
Product Details & Documents

Green screw cap, made of PP, for GL 45 threaded bottles and distributor body made of PP.

  • Very good chemical resistance
  • Temperature resistant from 0 to +110 °C
  • Suitable for vacuum applications

For the removal or filling of aggressive or pure liquids. For the connection of hoses, tubes and probes into vessels. Hoses with a maximum external diameter of  8,5 mm can be inserted through the necks.

Technische Daten
  • Rohr A-Ø:
    3 × max. 8,5 mm
  • Hals:
    3 × GL 14
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.