Septum caps, Nalgene®
38 - 430
Anbieter: Thermo Fisher Scientific
Direkt zu
Product Details & Documents
HDPE with silicone/PTFE septum.
- Sterile 10⁻⁶ SAL
- No adhesives used to attach septum to high density polyethylene closure
- Non-cytotoxic and non-pyrogenic
- Dual septum material for improved gas barrier
Technische Daten
- Verschlussgröße:38 - 430
- Cat. No.:NALG342178-0384
- Umweltzertifizierung:NAR Certificate of Retirement
- Umweltfreundlichkeit:Umweltschonende Herstellung