Standard ground joints
24/29 NS, - NS, 22 mm
Katalognr. QUTW1046
Anbieter: QUARTZTECHNISCHE WERKSTATTEN
Direkt zu
Product Details & Documents
Connections, quartz glass, with standard ground joint according to DIN 12 249.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion giving high resistance to temperature changes.
Technische Daten
- Kern:24/29 NS
- Hülse:- NS
- Rohr A-Ø:22 mm
- Seq:08