AuftragsverfolgungKontakt
Test kits, digital titration method
Mercuric nitrate digital titrator catridge, 2,570 N, 1
Katalognr. HACH2393701
Anbieter:   Hach
Test kits, digital titration method
Katalognr. HACH2393701
Anbieter:  Hach
CAS Nummer:  
Test kits, digital titration method
Test kits, digital titration method
Mercuric nitrate digital titrator catridge, 2,570 N, 1
CAS Nummer:  
Preis
70.10 SFr.
Einloggen für Verfügbarkeit
1 * 1 ST
Product Details & Documents
Digital titration provides a more precise and accurate method than the drop test method.
  • Reagents are pre-prepared
  • Exact comparison of colour
  • Rugged carrying case for analysis in the field
 : Please note some of the reagents supplied in the kits are hazardous, and appropriate precautions should be taken for use and disposal. Please contact Avantor for details.
Technische Daten
Technische Daten
  • VE:
    1
  • Bezeichnung:
    Mercuric nitrate digital titrator catridge, 2,570 N
Technische Daten