KIT,ABTS HRP SUBSTRATE,6X450ML 1 * 6 ST
KPLI50-62-01
Frequently Bought Together
VWR®, Mikroskop-Objektträger
: VWR Collection
VWR®, PCR-Gefäße, 0,2 ml
: VWR Collection
VWR®, Transferpipetten, High Performance
: VWR Collection
VWR®, Spreaders, L-shaped, Disposable
: VWR Collection
VWR®, Centrifuge Tubes, Conical Bottom
: VWR Collection
VWR®, Serologische Pipetten, Standard Serie
: VWR Collection
VWR®, Nitrilhandschuhe
: VWR Collection
VWR®, Transferpipetten, Standard Line
: VWR Collection
Safe-Lock Tubes, 2,0 ml - Microtubes
: EPPENDORF
Eppendorf Tubes® 3810X Microtubes 1,5 ml
: EPPENDORF
Safe-Lock Tubes, 1,5 ml - Microtubes
: EPPENDORF
Einwegspritzen, dreiteilig, HENKE-JECT®
: HENKE SASS WOLF