Bienenwachs
Glass bottle for solids, 250 g
Catalog # ROTH5830.1
Marketsource
Supplier: Roth Carl
Jump to:
Product Details & Documents
Synonyms: Wax, bees
- CAS Number:8012-89-3
- MDL Number:MFCD00132754
- Melting Pt:∼64 °C
- Density:0,95 g/cm³ (20 °C)
- Storage Temperature:Raumtemperatur
- EINECS:232-383-7
Specifications
Specifications
- Packung:Glass bottle for solids
- VE:250 g