Pipette tips, PreCision Line
Standard, 1000 µl, +, 1000 µl pipettes, 10 racks of 96 tips
Katalognr. BZYM761018
Marketsource
Anbieter: BIOZYM
Direkt zu
Product Details & Documents
Transparent, yellow or blue; sterile or non sterile.
- Free from DNase, RNase, ATP, pyrogens, additive, dispersant, wax and metals
- Autoclavable
- Produced under cleanroom conditions (VDI 2083 or class 1000/US-Fed. 209 H)
Zertifizierung: DIN 12650
Technische Daten
- Verpackt:10 racks of 96 tips
- Steril:+
- Volumen:1000 µl
- Seq:22
- Spitzentyp:Standard
- Zur Nutzung mit:1000 µl pipettes