Pipette tips, PreCision Line
Standard, 1000 µl, -, 1000 µl pipettes, 10 bags of 1000 tips
Katalognr. BZYM761011
Marketsource
Anbieter: BIOZYM
Direkt zu
Product Details & Documents
Transparent, yellow or blue; sterile or non sterile.
- Free from DNase, RNase, ATP, pyrogens, additive, dispersant, wax and metals
- Autoclavable
- Produced under cleanroom conditions (VDI 2083 or class 1000/US-Fed. 209 H)
Certifications: DIN 12650
Technische Daten
- Verpackt:10 bags of 1000 tips
- Steril:-
- Volumen:1000 µl
- Seq:19
- Spitzentyp:Standard
- Zur Nutzung mit:1000 µl pipettes