Pipette tips, PreCision Line
Blue, 1000 µl, -, 1000 µl pipettes, 50 bags of 1000 tips
BZYM761003
Marketsource
: BIOZYM
Transparent, yellow or blue; sterile or non sterile.
- Free from DNase, RNase, ATP, pyrogens, additive, dispersant, wax and metals
- Autoclavable
- Produced under cleanroom conditions (VDI 2083 or class 1000/US-Fed. 209 H)
: DIN 12650
- Verpackt:50 bags of 1000 tips
- Steril:-
- Volumen:1000 µl
- Seq:17
- Spitzentyp:Blue
- Zur Nutzung mit:1000 µl pipettes