Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Pipette tips, PreCision Line
Pipette tips, PreCision Line
Standard, 200 µl, -, 200 µl pipettes, 10 bags of 1000 tips
Catalog # BZYM760211
Marketsource
Supplier:   BIOZYM
Pipette tips, PreCision Line
Catalog # BZYM760211
Supplier:  BIOZYM
Marketsource
Pipette tips, PreCision Line
Standard, 200 µl, -, 200 µl pipettes, 10 bags of 1000 tips
Price
Inquire For Price
Log in for availability
1 * 10 ST
Product Details & Documents
Transparent, yellow or blue; sterile or non sterile.
  • Free from DNase, RNase, ATP, pyrogens, additive, dispersant, wax and metals
  • Autoclavable
  • Produced under cleanroom conditions (VDI 2083 or class 1000/US-Fed. 209 H)
Certifications: DIN 12650
Specifications
  • Verpackt:
    10 bags of 1000 tips
  • Steril:
    -
  • Volumen:
    200 µl
  • Seq:
    12
  • Spitzentyp:
    Standard
  • Zur Nutzung mit:
    200 µl pipettes