Pipette tips, PreCision Line
Yellow, 200 µl, -, 200 µl pipettes, 50 bags of 1000 tips
Katalognr. BZYM760203
Marketsource
Anbieter: BIOZYM
Direkt zu
Product Details & Documents
Transparent, yellow or blue; sterile or non sterile.
- Free from DNase, RNase, ATP, pyrogens, additive, dispersant, wax and metals
- Autoclavable
- Produced under cleanroom conditions (VDI 2083 or class 1000/US-Fed. 209 H)
Certifications: DIN 12650
Technische Daten
- Verpackt:50 bags of 1000 tips
- Steril:-
- Volumen:200 µl
- Seq:10
- Spitzentyp:Yellow
- Zur Nutzung mit:200 µl pipettes
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.