Pipette tips, PreCision Line
Crystal, 10 µl, -, 10 µl Eppendorf, Brand and Socorex pipettes, 10 racks of 96 tips
BZYM760005
Marketsource
: BIOZYM
Transparent, yellow or blue; sterile or non sterile.
- Free from DNase, RNase, ATP, pyrogens, additive, dispersant, wax and metals
- Autoclavable
- Produced under cleanroom conditions (VDI 2083 or class 1000/US-Fed. 209 H)
: DIN 12650
- Verpackt:10 racks of 96 tips
- Steril:-
- Volumen:10 µl
- Seq:03
- Spitzentyp:Crystal
- Zur Nutzung mit:10 µl Eppendorf, Brand and Socorex pipettes
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.