Pipette tips, PreCision Line
Crystal, 10 µl, -, 10 µl Eppendorf, Brand and Socorex pipettes, 10 bags of 1000 tips
Katalognr. BZYM760001
Marketsource
Anbieter: BIOZYM
Direkt zu
Product Details & Documents
Transparent, yellow or blue; sterile or non sterile.
- Free from DNase, RNase, ATP, pyrogens, additive, dispersant, wax and metals
- Autoclavable
- Produced under cleanroom conditions (VDI 2083 or class 1000/US-Fed. 209 H)
: DIN 12650
Technische Daten
- Verpackt:10 bags of 1000 tips
- Steril:-
- Volumen:10 µl
- Seq:01
- Spitzentyp:Crystal
- Zur Nutzung mit:10 µl Eppendorf, Brand and Socorex pipettes