AuftragsverfolgungKontakt
Syringes, SampleLock™ | Avantor
Syringes, SampleLock™
22, 50000 µl, 51 mm, 2, 1050 RN
Catalog # HAMI86336
Supplier:   HAMILTON BONADUZ
Syringes, SampleLock™
Catalog # HAMI86336
Supplier:  HAMILTON BONADUZ
Syringes, SampleLock™
Syringes, SampleLock™
22, 50000 µl, 51 mm, 2, 1050 RN
Price
SFr. 976.00
Log in for availability
1 * 1 ST
Product Details & Documents
These syringes allow the collection, storage, transport and analysis of liquid or gaseous samples without the danger of evaporative loss or environmental contamination.
  • Not autoclavable, gas sterilisable
  • An easy-to-use twist valve
  • A positive rear stop prevents loss of sample and plunger blowout
  • Fluid path includes SST, borosilicate glass and PTFE
The sample lock termination incorporates an on/off syringe valve with removable needle. This termination is used for headspace, environmental sample collection and storage, pre-pressurization of gaseous samples for GC analysis, and sample spiking.
Optional male luer and female luer lock adapters (sold separately) threads onto the SampleLock™ valve, making the syringes compatible with a multitude of connectors and fittings.
Specifications
  • Gauge:
    22
  • Typ:
    1050 RN
  • Spitzentyp:
    2
  • Volumen:
    50000 µl
  • Länge:
    51 mm
  • Seq:
    06
Accessories
Product Information
  • Catalog #
    Bezeichnung
    Availability
    Price
  • HAMI35081
    Removable needle adapter, to convert a large hub removable needle termination into a Female Luer Lock termination
    Log in for availability
    1 * 1 ST - SFr. 65.70
Product Information
  • Catalog #
    Bezeichnung
    Availability
    Price
  • HAMI35080
    Removable needle adapter, to convert a large hub removable needle termination into a Male Luer Tip (LT) termination
    Log in for availability
    1 * 1 ST - SFr. 53.80
Product Information
  • Catalog #
    Bezeichnung
    Availability
    Price
  • HAMI35083
    Removable needle adapter, to convert a large hub removable needle termination into a Male Luer Lock (TLL) termination
    Log in for availability
    1 * 1 ST - SFr. 58.50
Product Information
  • Catalog #
    Gauge
    Länge
    Ext.Ø
    Availability
    Price
  • HAMI7784-02
    22
    51 mm
    0,72 mm
    Log in for availability
    1 * 6 ST - SFr. 267.00
Product Information
  • Catalog #
    Gauge
    Länge
    Ext.Ø
    Availability
    Price
  • HAMI7779-01
    22
    51 mm
    0,72 mm
    Log in for availability
    1 * 6 ST - SFr. 92.00
Product Information
  • Catalog #
    Gauge
    Länge
    Ext.Ø
    Availability
    Price
  • HAMI7784-01
    22s
    51 mm
    0,72 mm
    Log in for availability
    1 * 6 ST - SFr. 271.00
Product Information
  • Catalog #
    Gauge
    Länge
    Ext.Ø
    Availability
    Price
  • HAMI7779-03
    22s
    51 mm
    0,72 mm
    Log in for availability
    1 * 6 ST - SFr. 92.00
Product Information
  • Catalog #
    Gauge
    Länge
    Ext.Ø
    Availability
    Price
  • HAMI7780-03
    22s
    51 mm
    0,72 mm
    Log in for availability
    1 * 6 ST - SFr. 72.90
Product Information
  • Catalog #
    Gauge
    Länge
    Ext.Ø
    Availability
    Price
  • HAMI7780-04
    22
    51 mm
    0,72 mm
    Log in for availability
    1 * 6 ST - SFr. 70.50
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.