Super syringes
Super syringe with TLL connector, 0,5 l, S0500 TLL
Katalognr. HAMI86311
Anbieter: HAMILTON BONADUZ
Direkt zu
Product Details & Documents
Super syringes with PTFE Luer lock (TLL) connector or tracheal adapter.
- Clear acrylic syringe barrels
- For use with gases and liquids
Ideal for use with air sampling, preparing gas standards, calibrating reservoirs, and pneumographs.
Technische Daten
- Typ:S0500 TLL
- Volumen:0,5 l
- Bezeichnung:Super syringe with TLL connector
Accessories
Produktinformationen
- Katalognr.BezeichnungFürVerfügbarkeitPreis
- HAMI18026TLL connectorsuper syringe S0500, S1000, S1500, S2000Einloggen für Verfügbarkeit1 * 1 ST - 94.40 SFr.