Deckgläser, rund
Round cover slides, 24 mm, Borosilikatglas
631-0672
Round cover glasses made of pure white borosilicate D 263 ™ M glass, Hydrolytic Class I.
- No bubbles or flaws
Suitable for in vitro diagnostic (IVD) applications.
: 100 cover glasses per box with separate lid.
- Ø:24 mm
- Bezeichnung:Round cover slides
- Material:Borosilikatglas
- Seq:010