Deckgläser, rund
Round cover slides, 16 mm, Borosilikatglas
Katalognr. 631-0668
Anbieter: MARIENFELD
Direkt zu
Product Details & Documents
Round cover glasses made of pure white borosilicate D 263 ™ M glass, Hydrolytic Class I.
- No bubbles or flaws
Suitable for in vitro diagnostic (IVD) applications.
: 100 cover glasses per box with separate lid.
Technische Daten
- Ø:16 mm
- Bezeichnung:Round cover slides
- Material:Borosilikatglas
- Seq:006
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.