Test tubes with rim
5 ml, 86 mm, 12 mm
Katalognr. 212-0183
Anbieter: KIMA SAS DI CHIARIN RENZO
Direkt zu
Product Details & Documents
PP, translucent or PS, clear, with rim.
- PS: Good thermal and chemical resistance
- PP: Excellent thermal and chemical resistance, autoclavable
- Robust, economical
Universal tubes for a wide range of applications.
Technische Daten
- Ext.Ø:12 mm
- Kapazität:5 ml
- Länge:86 mm