Deckgläser, rund
Round cover slides, 18 mm, Borosilikatglas
Katalognr. 631-0669
Anbieter: MARIENFELD
Direkt zu
Product Details & Documents
Round cover glasses made of pure white borosilicate D 263 ™ M glass, Hydrolytic Class I.
- No bubbles or flaws
Suitable for in vitro diagnostic (IVD) applications.
Verpackung: 100 cover glasses per box with separate lid.
Technische Daten
- Ø:18 mm
- Bezeichnung:Round cover slides
- Material:Borosilikatglas
- Seq:007