AuftragsverfolgungKontakt
Protein G
Protein G
Protein G peroxidase conjugated, 1 mg
Catalog # ROCKPG00-03
Protein G
Catalog # ROCKPG00-03
CAS Number:  
Protein G
Protein G
Protein G peroxidase conjugated, 1 mg
CAS Number:  
Price
SFr. 685.00
Log in for availability
1 * 1 mg
Product Details & Documents
Protein G is a surface protein of two groups of streptococcal bacteria that has ability to bind immunoglobulins and also to albumins. Protein G are prepared from chromatographically pure protein G.
Protein G is stable at 4 °C as an undiluted liquid. Freezing alkaline phosphatase conjugates will result in a substantial loss of enzymatic activity.
Protein G alkalline phosphatase conjugated is ideal for signal transduction, molecular biology, cancer, and cell signaling. Protein G peroxidase conjugated is a useful reagent in western blotting and ELISA experiments. Protein G fluorescein conjugated is suitable for immunomicroscopy and flow cytometry or FACS analysis.
Documents
Specifications
Specifications
  • VE:
    1 mg
  • Bezeichnung:
    Protein G peroxidase conjugated