Human plasma
Human plasma in sodium heparin, sterile, 50 mL
Katalognr. ROCKD519-06-0050
Anbieter: Rockland Immunochemicals
Direkt zu
Product Details & Documents
Plasma is obtained from non-hemolysed blood collected from healthy, fasted donors. Sterilisation is accomplished by filtration.
Documents
D519-03-0050_ D519-04-0050_
Fiche technique
Technische Daten
Technische Daten
- VE:50 mL
- Bezeichnung:Human plasma in sodium heparin, sterile
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.