Human plasma
Human plasma in sodium EDTA, sterile, 1.000 mL
Katalognr. ROCKD519-04-1000
Anbieter: Rockland Immunochemicals
Direkt zu
Product Details & Documents
Plasma is obtained from non-hemolysed blood collected from healthy, fasted donors. Sterilisation is accomplished by filtration.
Documents
D519-03-0050_ D519-04-0050_
Datasheet
Technische Daten
Technische Daten
- VE:1.000 mL
- Bezeichnung:Human plasma in sodium EDTA, sterile