Human plasma
Human plasma in sodium citrate, sterile, 500 mL
ROCKD519-03-0500
Plasma is obtained from non-hemolysed blood collected from healthy, fasted donors. Sterilisation is accomplished by filtration.
- VE:500 mL
- Bezeichnung:Human plasma in sodium citrate, sterile