Human plasma
Human plasma in sodium citrate, sterile, 50 mL
Katalognr. ROCKD519-03-0050
Anbieter: Rockland Immunochemicals
Direkt zu
Product Details & Documents
Plasma is obtained from non-hemolysed blood collected from healthy, fasted donors. Sterilisation is accomplished by filtration.
Technische Daten
Technische Daten
- VE:50 mL
- Bezeichnung:Human plasma in sodium citrate, sterile