AuftragsverfolgungKontakt
Human plasma
Human plasma in potassium EDTA, sterile, 50 mL
Catalog # ROCKD519-05-0050
Human plasma
Catalog # ROCKD519-05-0050
CAS Number:  
Human plasma
Human plasma
Human plasma in potassium EDTA, sterile, 50 mL
CAS Number:  
Price
SFr. 1’390.00
Log in for availability
1 * 50 mL
Product Details & Documents
Plasma is obtained from non-hemolysed blood collected from healthy, fasted donors. Sterilisation is accomplished by filtration.
Specifications
Specifications
  • VE:
    50 mL
  • Bezeichnung:
    Human plasma in potassium EDTA, sterile