Human plasma
Human plasma in potassium EDTA, sterile, 50 mL
Catalog # ROCKD519-05-0050
Supplier: Rockland Immunochemicals
Jump to:
Product Details & Documents
Plasma is obtained from non-hemolysed blood collected from healthy, fasted donors. Sterilisation is accomplished by filtration.
Specifications
Specifications
- VE:50 mL
- Bezeichnung:Human plasma in potassium EDTA, sterile
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.