Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Human plasma
Human plasma
Human plasma in alsevers, sterile, 50 mL
Katalognr. ROCKD519-02-0050
Human plasma
Katalognr. ROCKD519-02-0050
CAS Nummer:  
Human plasma
Human plasma
Human plasma in alsevers, sterile, 50 mL
CAS Nummer:  
Preis
SFr. 1’390.00
Einloggen für Verfügbarkeit
1 * 50 mL
Product Details & Documents
Plasma is obtained from non-hemolysed blood collected from healthy, fasted donors. Sterilisation is accomplished by filtration.
Documents
Technische Daten
Technische Daten
  • VE:
    50 mL
  • Bezeichnung:
    Human plasma in alsevers, sterile
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.