Grounding cables, Super Erecta®
Grounding cable with spring-loaded clamp attachment
Katalognr. METOASK16S
Anbieter: Metro International
Direkt zu
Product Details & Documents
These grounding cables provide an effective method of electrostatic discharge for any Metro Super Erecta unit (stationary or mobile) when used in conjunction with aluminum split sleeves or ESD kit and conductive casters.
- Cables ensure a proper path to ground when used with ESD flooring
- Grounding cable with universal bracket fits wire and solid shelves, as well as open-wire frames
Technische Daten
- Bezeichnung:Grounding cable with spring-loaded clamp attachment
- Seq:01