n-Dodecyl-β-D-maltosid
Glass bottle for solids, 1 g
Katalognr. BMOL56366-1
Anbieter: Biomol Enzo
Direkt zu
Product Details & Documents
Synonyme: DDM, Lauryl-β-D-maltoside, Dodecyl-β-D-maltopyranoside, (2R,3R,4S,5S,6R)-2-(((2R,3S,4R,5R,6R)-6-(Dodecyloxy)-4,5-dihydroxy-2-(hydroxymethyl)tetrahydro-2H-pyran-3-yl)oxy)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol, N-Dodecyl-Beta-Maltoside
- CAS Nummer:69227-93-6
- Formel:C₂₄H₄₆O₁₁
- MDL-Nummer:MFCD00043012
- Molekulargewicht:510.6 g/mol
- Lagertemperatur:Gefrierschrank
Technische Daten
Technische Daten
- Packung:Glass bottle for solids
- VE:1 g



