Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
(R)-(-)-2-Aminohexan ≥99%, ee 96%, ChiPros®

(R)-(-)-2-Aminohexan ≥99%, ee 96%, ChiPros®

Glasflasche für Feststoffe, 25 g
Catalog # L19304.14
Flammable
Corrosive
Irritant
(R)-(-)-2-Aminohexan ≥99%, ee 96%, ChiPros®
Catalog # L19304.14
CAS Number:  70095-40-8
(R)-(-)-2-Aminohexan ≥99%, ee 96%, ChiPros®
(R)-(-)-2-Aminohexan ≥99%, ee 96%, ChiPros®

Glasflasche für Feststoffe, 25 g

CAS Number:  70095-40-8
Price
SFr. 855.00
Log in for availability
1 * 25 g
Product Details & Documents

Purity: 99+% CAS No.: 70095-40-8 Molecular Formula: C6H15N Formula Weight: 101.19 Flash Point: -3°(26°F) Melting Point: -20° Boiling Point: 116° Beilstein Reference: 1718994 UN No.: 2733 MDL No.: MFCD00671627

  • CAS Number:70095-40-8
  • Formula:C₆H₁₅N
  • MDL Number:MFCD00671627
  • Molecular Weight:101.19 g/mol
  • Boiling Pt:116 °C
  • Melting Pt:< –20 °C
  • Density:0,755
  • Flash Pt:–3 °C (26 °F)
  • Storage Temperature:Raumtemperatur
  • UN:2733
  • ADR:3,II
Specifications

Specifications

  • Packung:
    Glasflasche für Feststoffe
  • VE:
    25 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.