(R)-(-)-2-Aminohexan ≥99%, ee 96%, ChiPros®
Glasflasche für Feststoffe, 1 g
Anbieter: Thermo Fisher Scientific
(R)-(-)-2-Aminohexan ≥99%, ee 96%, ChiPros®
Katalognr. L19304.03
Anbieter: Thermo Fisher Scientific
CAS Nummer: 70095-40-8
Direkt zu
Product Details & Documents
Purity: 99+% CAS No.: 70095-40-8 Molecular Formula: C6H15N Formula Weight: 101.19 Flash Point: -3°(26°F) Melting Point: -20° Boiling Point: 116° Beilstein Reference: 1718994 UN No.: 2733 MDL No.: MFCD00671627
- CAS Nummer:70095-40-8
- Formel:C₆H₁₅N
- MDL-Nummer:MFCD00671627
- Molekulargewicht:101.19 g/mol
- Siedepunkt:116 °C
- Schmelzender Platingehalt:< –20 °C
- Dichte:0,755
- Blitz Pt:–3 °C (26 °F)
- Lagertemperatur:Raumtemperatur
- UN:2733
- ADR:3,II
Documents
Technische Daten
Technische Daten
- Packung:Glasflasche für Feststoffe
- VE:1 g
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



