Methanol ≥99%
Glasflasche, 1.000 mL
Anbieter: Thermo Fisher Scientific
Direkt zu
Product Details & Documents
Purity: 99% CAS No.: 67-56-1 Molecular Formula: CH3OH Formula Weight: 32.04 Flash Point: 11°(52°F) Refractive Index: 1.3290 Odour: Mild Melting Point: -98° Boiling Point: 64.7° Beilstein Reference: 1098229 Merck Reference: 14,5957 UN No.: 1230 MDL No.: MFCD00004595
Synonyme: Wood alcohol, Methyl alcohol, Hydroxy methane, Carbinol
- CAS Nummer:67-56-1
- Formel:CH₃OH
- MDL-Nummer:MFCD00004595
- Molekulargewicht:32.04 g/mol
- Merck-Index:14,5957
- Siedepunkt:64,7 °C
- Schmelzender Platingehalt:–98 °C
- Dichte:0,791
- Blitz Pt:11 °C (52 °F)
- Lagertemperatur:Raumtemperatur
- EINECS:200-659-6
- UN:1230
- ADR:3,II
- ERREICHEN:01-2119433307-44
Technische Daten
Technische Daten
- Packung:Glasflasche
- VE:1.000 mL



