Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Sampling cannulae

Sampling cannulae

Bent cannula with Luer lock, PEEK, 3,2×120 mm
Catalog # QLAACAN475-PH
Sampling cannulae
Catalog # QLAACAN475-PH
Sampling cannulae
Sampling cannulae

Bent cannula with Luer lock, PEEK, 3,2×120 mm

Price
SFr. 259.00
Log in for availability
1 * 1 ST
Product Details & Documents

Standard cannula are unique, proprietary configurations for most manual and automated sampling applications.

  • Manufactured in both electropolished 316 stainless steel or PEEK
  • Can accommodate all standard tip filters
  • Optional stoppers set the proper sampling position

The USP defines the sampling point as: 'A zone half way between the top of the media and top of the paddle or basket not less than 1 cm from the vessel wall. The sampling position is critical but so too is the type of sampling cannula (also known as a probe) used.'

Specifications
  • Ø×L:
    3,2×120 mm
  • Bezeichnung:
    Bent cannula with Luer lock
  • Material:
    PEEK
  • Seq:
    201
Accessories

Product Information

  • Catalog #
    Bezeichnung
    Availability
    Price
  • QLAACANSTP-HR
    Stopper for adjustable cannula, Hanson, Ø = 3,2 mm
    Log in for availability
    1 * 1 ST

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.