Ethylacetat ≥99%
Glasflasche, 2.500 mL
Anbieter: Thermo Fisher Scientific
Direkt zu
Product Details & Documents
Purity: 99% CAS No.: 141-78-6 Molecular Formula: C4H8O2 Formula Weight: 88.11 Flash Point: -3°(26°F) Refractive Index: 1.3720 Odour: Sweet Melting Point: -84° Boiling Point: 76-77° Beilstein Reference: 506104 Merck Reference: 14,3757 UN No.: 1173 MDL No.: MFCD00009171
Synonyme: Acetic acid ethyl ester
- CAS Nummer:141-78-6
- Formel:CH₃COOC₂H₅
- MDL-Nummer:MFCD00009171
- Molekulargewicht:88.11 g/mol
- Merck-Index:14,3757
- Siedepunkt:76…77 °C
- Schmelzender Platingehalt:–84 °C
- Dichte:0,902
- Blitz Pt:–3 °C (26 °F)
- Lagertemperatur:Raumtemperatur
- EINECS:205-500-4
- UN:1173
- ADR:3,II
- ERREICHEN:01-2119475103-46
Technische Daten
Technische Daten
- Packung:Glasflasche
- VE:2.500 mL
- Cat. No.:L10925.0F
- Umweltzertifizierung:Chem21 Solvent Selection Guide recommended by default and after discussion
- Umweltfreundlichkeit:Unbedenklicher für Mensch und Umwelt
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



