Bond Elut adapters
Adapter cap for 60 ml Bond Elut cartridges
Katalognr. VARI12131004
Anbieter: VARIAN
Direkt zu
Product Details & Documents
Bond Elut adapters fit on top of any Bond Elut cartridge and contain a female Luer fitting that accommodates the tip of another cartridge.
- Connect SPE cartridges in series for large samples
- Expand cartridge volume for even more applications
- Transfer large-volume samples to any SPE cartridge
Technische Daten
- Bezeichnung:Adapter cap for 60 ml Bond Elut cartridges
- Seq:02