Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Water testing kit, MINIKIT
Water testing kit, MINIKIT
Sulphite (high range), 20 - 500 mg/l Na2SO3, AF 435, 1 KIT
Katalognr. 705-0421
Anbieter:   Lovibond
Water testing kit, MINIKIT
Katalognr. 705-0421
Anbieter:  Lovibond
CAS Nummer:  
Water testing kit, MINIKIT
Water testing kit, MINIKIT
Sulphite (high range), 20 - 500 mg/l Na2SO3, AF 435, 1 KIT
CAS Nummer:  
Preis
91.60 SFr.
Einloggen für Verfügbarkeit
1 * 1 KIT
Product Details & Documents
MINIKIT provides quick and simple tests for the analysis of boiler water, cooling water, drinking and industrial water.
  • MINIKITS are based on titrimetric methods
  • Easy operation and exact reagent dosing
  • Foil-wrapped tablet reagents with a minimum guaranteed shelf life of five years
Tablet count method: In this method the liquid titration solution and indicator are replaced by AQUALYTIC® tablet reagents. A specific number of tablets is added to a defined sample volume until a chemically induced colour change takes place. The concentration of the parameter being measured is calculated from the number of tablets required. The measuring range can be expanded by varying the sample volume.
Yes/No test: This method tells whether a specific ingredient is present in the water and/or if its concentration is higher or lower than a defined level.
Turbidity method: A two-section calibrated test tube is filled with the water sample and a reagent tablet added. The reagent creates a level of turbidity that is proportional to the concentration of the parameter being measured. The inner tube, which has a black dot on its base, is lowered until the dot is obscured by the turbidity. The result is read off from the water level in the inner tube.
Documents
Technische Daten
Technische Daten
  • VE:
    1 KIT
  • Bezeichnung:
    Sulphite (high range)
  • Typ:
    AF 435
  • Messbereich:
    20 - 500 mg/l Na2SO<sub>3</sub>
Technische Daten
Accessories
Produktinformationen
  • Katalognr.
    Bezeichnung
    Verfügbarkeit
    Preis
  • LOVI385130
    Gefäß zum Verdünnen/Vermischen mit Deckel, Kunststoff, graduiert, 100 ml
    Einloggen für Verfügbarkeit
    1 * 1 ST - SFr. 14.20
Produktinformationen
  • Katalognr.
    Bezeichnung
    Verfügbarkeit
    Preis
  • LOVI385132
    Gefäß zum Verdünnen/Vermischen mit Deckel, Kunststoff, graduiert, 30 ml
    Einloggen für Verfügbarkeit
    1 * 1 ST - SFr. 14.20
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.