Measuring Cylinders
10 ml, 0,2 ml, 140 mm, 0,2 ± ml
Katalognr. 612-0057
Anbieter: SciLabware
Direkt zu
Product Details & Documents
Borosilicate glass, tall form, class B.
- With spout
- Graduations in high contrast white enamel
- Hexagonal bases on cylinders above 5 ml for greater stability
Zertifizierung: BS 604, ISO 4788, DIN 12 680
Dokumente
Technische Daten
- Kapazität:10 ml
- Höhe:140 mm
- Teilung:0,2 ml
- Toleranz:0,2 ± ml