Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
 

Natriumhydroxid, Supelco®

Wellpappfaltkisten, 50 kg
  1.06467.9050
 
 :   Merck
Natriumhydroxid, Supelco®
  1.06467.9050
 :  Merck
 :  1310-73-2
Natriumhydroxid, Supelco®

Wellpappfaltkisten, 50 kg

 :  1310-73-2
 
SFr. 1’010.00
   
1 * 50 kg
 
 :  Caustic soda
  •  :1310-73-2
  •  :NaOH
  •  :MFCD00003548
  •  :40 g/mol
  •  :14,08627
  •  :1390 °C (1013 hPa)
  •  :323 °C
  •  :2,13 g/cm³ (20 °C)
  •  :215-185-5
  •  :8,II
  •  :M-Clarity™ Qualitätsstufe = MQ200
  •  :Raumtemperatur
  •  :1823
  •  :01-2119457892-27
 

 

  • Solubility:
    1090 g/l (20 °C)
  • Melting Point:
    323 °C
  • Molar Mass:
    40.00 g/mol
  • Boiling Point:
    1390 °C (1013 hPa)
  • Vapor Pressure:
    (20 °C)
  • Density:
    2.13 g/cm3 (20 °C)
  • pH:
    14 (50 g/l, H2O, 20 °C)
  • Hazard Class:
    8
  • Hazard Class Description:
    Corrosive Material

 

  • Packung:
    Wellpappfaltkisten
  • VE:
    50 kg

 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.