Combustion boats
50×15×9 mm
Katalognr. TSLA73375
Anbieter: TECHNICAL QUARTZ SOLUTIONS
Direkt zu
Product Details & Documents
Combustion boats, quartz, glazed, with or without handle.
- Very good chemical resistance
- Very good temperature resistance
Technische Daten
- Seq:01
- B×T×H:50×15×9 mm