Tropin ≥98%
Glass bottle for solids, 10 g
Anbieter: Thermo Fisher Scientific
Direkt zu
Product Details & Documents
Purity: 98% CAS No.: 120-29-6 Molecular Formula: C8H15NO Formula Weight: 141.22 Storage Temperature: 2°C to 8°C Odour: Amine-like pH: 12 Melting Point: 61-65° (dried) Boiling Point: 232-234° Beilstein Reference: 80188 Merck Reference: 14,9781 MDL No.: MFCD00005551
Synonyme: endo-8-Methyl-8-azabicyclo[3.2.1]octan-3-ol
- CAS Nummer:120-29-6
- Formel:C₈H₁₅NO
- MDL-Nummer:MFCD00180667
- Molekulargewicht:141.21 g/mol
- Merck-Index:14,9781
- Siedepunkt:232…234 °C
- Schmelzender Platingehalt:61…65 °C (wasserfrei)
- Dichte:1,04
- EINECS:204-384-2
Documents
Technische Daten
Technische Daten
- Packung:Glass bottle for solids
- VE:10 g



