Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Test Tubes with Rim, Medium Wall | Avantor

Test Tubes with Rim, Medium Wall

150 mm, 24 mm
Katalognr. 212-7408
Anbieter:   SciLabware
Test Tubes with Rim, Medium Wall
Katalognr. 212-7408
Anbieter:  SciLabware
Test Tubes with Rim, Medium Wall
Test Tubes with Rim, Medium Wall

150 mm, 24 mm

Preis
SFr. 279.00
Einloggen für Verfügbarkeit
1 * 100 ST
Product Details & Documents

PYREX®, borosilicate glass.

  • Superior performance in comparison to neutral borosilicate and soda-lime glass
  • Excellent resistance to chemicals and high temperature

High quality test tubes designed and constructed for optimum performance.

 : ISO 4142

Technische Daten
  • Ext.Ø:
    24 mm
  • Länge:
    150 mm
  • Seq:
    09