Morpholin ≥99%
Glasflasche für Feststoffe, 100 g
Anbieter: Thermo Fisher Scientific
Direkt zu
Product Details & Documents
Purity: 99% CAS No.: 110-91-8 Molecular Formula: C4H9NO Formula Weight: 87.12 Flash Point: 35°(95°F) Physical Form: Liquid Density: 1.007 Refractive Index: 1.4540 Sensitivity: Hygroscopic Appearance: Colourless Odour: Amine-like Melting Point: -5° Boiling Point: 129° Beilstein Reference: 102549 Merck Reference: 14,6277 UN No.: 2054 MDL No.: MFCD00005972
- CAS Nummer:110-91-8
- Formel:C₄H₉NO
- MDL-Nummer:MFCD00005972
- Molekulargewicht:87.12 g/mol
- Merck-Index:14,6277
- Siedepunkt:129 °C
- Schmelzender Platingehalt:–5 °C
- Dichte:1,007
- Blitz Pt:35 °C (95 °F)
- Lagertemperatur:Raumtemperatur
- EINECS:203-815-1
- UN:2054
- ADR:8,I
Documents
Technische Daten
Technische Daten
- Packung:Glasflasche für Feststoffe
- VE:100 g
- Cat. No.:A10355.22
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



