Sulfolan ≥99%
Glasflasche für Feststoffe, 1.000 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Purity: 99% CAS No.: 126-33-0 Molecular Formula: C4H8O2S Formula Weight: 120.17 Flash Point: 166°(330°F) Refractive Index: 1.4825 Odour: Weak, characteristic Melting Point: 25-28° Boiling Point: 285° Beilstein Reference: 107765 Merck Reference: 14,8955 MDL No.: MFCD00005484
Synonyms: Tetrahydrothiophene-1,1-dioxide, Tetramethylene sulphone, Tetrahydrothiophene 1,1-dioxide
- CAS Number:126-33-0
- Formula:C₄H₈O₂S
- MDL Number:MFCD00005484
- Molecular Weight:120.17 g/mol
- Merck Index:14,8955
- Boiling Pt:285 °C
- Melting Pt:25…28 °C
- Density:1,261
- Flash Pt:166 °C (330 °F)
- Storage Temperature:Raumtemperatur
- EINECS:204-783-1
Documents
Certificates
Search by Lot NumberOrganic Building Blocks and Intermediates
Application Notes
Thermo Scientific Stockroom Essentials
Product Literature
Specifications
Specifications
- Packung:Glasflasche für Feststoffe
- VE:1.000 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



