Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
 

Notfallbeutel-Kit, PIG®

Spill kit, includes 4 absorbent socks, 28 mat pads and 5 disposal bags, 34,7 L, 150×430×520 mm
  121-0515
 :   NEW PIG
Notfallbeutel-Kit, PIG®
  121-0515
 :  NEW PIG
Notfallbeutel-Kit, PIG®
Notfallbeutel-Kit, PIG®

Spill kit, includes 4 absorbent socks, 28 mat pads and 5 disposal bags, 34,7 L, 150×430×520 mm

 
SFr. 217.00
   
1 * 1 KIT
 

PIG absorbents in the ideal spill kit bag - hi-viz, water-resistant, and easy to close, hang and carry.

  • Durable, water-resistant bag with hook-and-loop closure keeps contents clean and dry
  • PIG aborbent socks stop spreading spills; PIG mat absorbs quickly
  • Temporary disposal bags help make cleanup easier
  • Bright-yellow bag with bold Spill Kit lettering for quick identification
  • Strap for easy carrying and hanging

Compact and easy to store kit is great for use in trucks or equipment.

 : To ensure effectiveness and your safety, we recommend that you conduct compatibility and absorption testing of your chemicals with PIG® HAZ-MAT products prior to purchase.

 
  • Bezeichnung:
    Spill kit, includes 4 absorbent socks, 28 mat pads and 5 disposal bags
  • Seq:
    02
  • B×T×H:
    150×430×520 mm
  • Absorptionsvermögen:
    34,7 L
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.