Measuring Cylinders
2000 ml, 20 ml, 570 mm, 20 ± ml
Katalognr. 612-0064
Anbieter: SciLabware
Direkt zu
Product Details & Documents
Borosilicate glass, tall form, class B.
- With spout
- Graduations in high contrast white enamel
- Hexagonal bases on cylinders above 5 ml for greater stability
Zertifizierung: BS 604, ISO 4788, DIN 12 680
Dokumente
Technische Daten
- Kapazität:2000 ml
- Höhe:570 mm
- Teilung:20 ml
- Toleranz:20 ± ml